The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007620.1 | NP_001007621.1 | 5 - 6 | 632 | 117 |
XM_006251754.1 | XP_006251816.1 | 6 - 7 | 492 | 117 | |
GenBank mRNA | BC079137.1 | - | 5 - 6 | 632 | 117 |
FQ212778.1 | - | 5 - 6 | 117 | ||
FQ214416.1 | - | 5 - 6 | 321 | 117 | |
FQ234205.1 | - | 5 - 6 | 321 | 117 |