Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fto
Entrez Gene ID
291905
Gene Name
fat mass and obesity associated
Gene Aliases
RGD1305121
Chromosome Location
Chr.19: 16734271 - 17115098 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039713.1 NP_001034802.1 7 - 8 1230 98
XM_006255176.2 XP_006255238.1 7 - 8 1663 98
XM_008772284.1 XP_008770506.1 7 - 8 1559 98
XM_008772285.1 XP_008770507.1 7 - 8 1561 98
GenBank mRNA AM233906.1 - 7 - 8 1230 98
BC168239.1 - 7 - 8 1245 98
FQ211940.1 - 7 - 8 1270 98
FQ212297.1 - 7 - 8 1259 98

View Full Product Details