Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Papd5
Entrez Gene ID
307745
Gene Name
PAP associated domain containing 5
Gene Aliases
-
Chromosome Location
Chr.19: 19771518 - 19834837 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107416.1 NP_001100886.1 2 - 3 339 112

View Full Product Details