Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ryr1
Entrez Gene ID
114207
Gene Name
ryanodine receptor 1 (skeletal)
Gene Aliases
RYR-1, Ryr1l
Chromosome Location
Chr.1: 87959596 - 88066248 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008759293.1 XP_008757515.1 91 - 92 13033 88

View Full Product Details