The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012523.2 | NP_036655.1 | 7 - 8 | 733 | 97 |
GenBank mRNA | BC078900.1 | - | 7 - 8 | 684 | 97 |
M57276.1 | - | 7 - 8 | 747 | 97 |