Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tnnt1
Entrez Gene ID
171409
Gene Name
troponin T type 1 (skeletal, slow)
Gene Aliases
Fang2, sTnT, tnTs
Chromosome Location
Chr.1: 72889270 - 72899629 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134388.2 NP_599215.2 5 - 6 152 84
GenBank mRNA AF399874.1 - 4 - 5 100 84
AY334079.1 - 4 - 5 96 84

View Full Product Details