The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033653.1 | NP_001028825.1 | 2 - 3 | 4999 | 89 |
NM_057103.2 | NP_476444.2 | 3 - 4 | 5228 | 89 | |
XM_006227847.2 | XP_006227909.1 | 2 - 3 | 5490 | 89 | |
XM_006227848.2 | XP_006227910.1 | 2 - 3 | 5430 | 89 | |
GenBank mRNA | AY695056.1 | - | 3 - 4 | 5228 | 89 |
AY695057.1 | - | 2 - 3 | 4999 | 89 | |
AY695058.1 | - | 2 - 3 | 4953 | 89 | |
U41453.1 | - | 3 - 4 | 5103 | 89 |