Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lphn3
Entrez Gene ID
170641
Gene Name
latrophilin 3
Gene Aliases
Cirl3, Lec3
Chromosome Location
Chr.14: 28362176 - 28856419 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130822.1 NP_570835.1 17 - 18 3336 111
XM_008769968.1 XP_008768190.1 17 - 18 3440 111
XM_008769969.1 XP_008768191.1 17 - 18 3425 111
XM_008769970.1 XP_008768192.1 17 - 18 3440 111
XM_008769971.1 XP_008768193.1 17 - 18 3440 111
XM_008769972.1 XP_008768194.1 16 - 17 3401 111
XM_008769973.1 XP_008768195.1 17 - 18 3440 111
XM_008769974.1 XP_008768196.1 17 - 18 3440 111
XM_008769975.1 XP_008768197.1 17 - 18 3440 111
XM_008769976.1 XP_008768198.1 17 - 18 3440 111
XM_008769977.1 XP_008768199.1 16 - 17 3236 111
XM_008769978.1 XP_008768200.1 16 - 17 3401 111
XM_008769979.1 XP_008768201.1 16 - 17 3236 111
XM_008769980.1 XP_008768202.1 17 - 18 3440 111
XM_008769981.1 XP_008768203.1 17 - 18 3440 111
XM_008769982.1 XP_008768204.1 17 - 18 3439 111
XM_008769983.1 XP_008768205.1 17 - 18 3439 111
XM_008769984.1 XP_008768206.1 17 - 18 3439 111
XM_008769985.1 XP_008768207.1 16 - 17 3235 111
XM_008769986.1 XP_008768208.1 16 - 17 3235 111
GenBank mRNA AF063103.1 - 17 - 18 3336 111
AF081154.1 - 16 - 17 3157 111
AF081155.1 - 16 - 17 3157 111
AF081156.1 - 16 - 17 3157 111
AF081157.1 - 17 - 18 3361 111
AF081158.1 - 17 - 18 3361 111
AF081159.1 - 17 - 18 3361 111

View Full Product Details