The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139327.1 | NP_647543.1 | 3 - 4 | 271 | 87 |
XM_006256156.1 | XP_006256218.1 | 4 - 5 | 418 | 87 | |
XM_006256157.2 | XP_006256219.1 | 5 - 6 | 531 | 87 | |
XM_006256158.2 | XP_006256220.1 | 4 - 5 | 460 | 87 | |
XM_006256161.1 | XP_006256223.1 | 4 - 5 | 385 | 87 | |
XM_006256162.2 | XP_006256224.1 | 4 - 5 | 421 | 87 | |
XM_006256163.2 | XP_006256225.1 | 4 - 5 | 796 | 87 | |
GenBank mRNA | AF507966.1 | - | 3 - 4 | 271 | 87 |
AF511040.1 | - | 3 - 4 | 258 | 87 |