The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178333.1 | NP_848016.1 | 16 - 17 | 2117 | 97 |
XM_008773069.1 | XP_008771291.1 | 18 - 19 | 3447 | 97 | |
XM_008773070.1 | XP_008771292.1 | 18 - 19 | 3438 | 97 | |
GenBank mRNA | AB098162.1 | - | 16 - 17 | 2117 | 97 |