Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rfng
Entrez Gene ID
60433
Gene Name
RFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
Gene Aliases
-
Chromosome Location
Chr.10: 109963726 - 109966847 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021849.1 NP_068621.1 2 - 3 324 87
XM_006247910.2 XP_006247972.1 2 - 3 390 87
XM_008768480.1 XP_008766702.1 2 - 3 390 87
XM_008768481.1 XP_008766703.1 2 - 3 390 87
GenBank mRNA AB016486.1 - 2 - 3 324 87
BC062031.1 - 2 - 3 369 87

View Full Product Details