Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prpsap1
Entrez Gene ID
64390
Gene Name
phosphoribosyl pyrophosphate synthetase-associated protein 1
Gene Aliases
-
Chromosome Location
Chr.10: 105430516 - 105457942 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022545.3 NP_071990.2 1 - 2 337 126
GenBank mRNA BC078822.1 - 1 - 2 312 126
D26073.1 - 1 - 2 235 126

View Full Product Details