Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prx
Entrez Gene ID
78960
Gene Name
periaxin
Gene Aliases
-
Chromosome Location
Chr.1: 84302912 - 84324560 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023976.2 NP_076466.2 1 - 2 35 64
GenBank mRNA Z29649.2 - 1 - 2 35 64

View Full Product Details