Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Acsm4
Entrez Gene ID
353317
Gene Name
acyl-CoA synthetase medium-chain family member 4
Gene Aliases
Omacs
Chromosome Location
Chr.1: 189432604 - 189457052 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181695.1 NP_859046.1 12 - 13 1660 97
GenBank mRNA AB096688.1 - 12 - 13 1660 97

View Full Product Details