Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ldlrad3
- Entrez Gene ID
-
366138
- Gene Name
- low density lipoprotein receptor class A domain containing 3
- Gene Aliases
-
RGD1307846
- Chromosome Location
-
Chr.3: 91595448 - 91839054 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_345406.7 |
XP_345407.4 |
3 - 4 |
397 |
79 |
View Full Product Details