Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ism1
Entrez Gene ID
311760
Gene Name
isthmin 1, angiogenesis inhibitor
Gene Aliases
RGD1562551
Chromosome Location
Chr.3: 132736433 - 132808544 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134552.2 NP_001128024.1 - 367 66

View Full Product Details