Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tspan18
Entrez Gene ID
311210
Gene Name
tetraspanin 18
Gene Aliases
RGD1560411
Chromosome Location
Chr.3: 82097068 - 82236692 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107750.1 NP_001101220.1 1 - 2 334 119

View Full Product Details