The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107750.1 | NP_001101220.1 | 9 - 10 | 1199 | 68 |
XM_006234573.2 | XP_006234635.1 | 9 - 10 | 1130 | 68 | |
XM_006234574.2 | XP_006234636.1 | 10 - 11 | 1272 | 68 | |
XM_006234575.2 | XP_006234637.1 | 8 - 9 | 1050 | 68 | |
XM_008762027.1 | XP_008760249.1 | 8 - 9 | 950 | 68 |