Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Glcci1
Entrez Gene ID
296884
Gene Name
glucocorticoid induced transcript 1
Gene Aliases
RGD1563612
Chromosome Location
Chr.4: 34282625 - 34612709 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177442.1 NP_001170913.1 8 - 9 964 94

View Full Product Details