Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc35a3
Entrez Gene ID
310808
Gene Name
solute carrier family 35 (UDP-N-acetylglucosamine (UDP-GlcNAc) transporter), member A3
Gene Aliases
-
Chromosome Location
Chr.2: 219702681 - 219741886 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012082.1 NP_001012082.1 4 - 5 604 84
XM_008761461.1 XP_008759683.1 4 - 5 872 84
GenBank mRNA BC079371.1 - 4 - 5 604 84

View Full Product Details