Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc39a8
Entrez Gene ID
295455
Gene Name
solute carrier family 39 (zinc transporter), member 8
Gene Aliases
-
Chromosome Location
Chr.2: 241029188 - 241092584 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011952.1 NP_001011952.1 2 - 3 644 73
XM_006233356.2 XP_006233418.1 3 - 4 683 73
XM_006233357.2 XP_006233419.1 3 - 4 903 73
GenBank mRNA BC089844.1 - 2 - 3 644 73

View Full Product Details