Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr77
Entrez Gene ID
310769
Gene Name
WD repeat domain 77
Gene Aliases
RGD1310479
Chromosome Location
Chr.2: 208577292 - 208587263 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008771.3 NP_001008771.2 9 - 10 960 60
GenBank mRNA AY321348.1 - 10 - 11 921 60
BC097367.1 - 9 - 10 911 60
FQ213175.1 - 9 - 10 1055 60
FQ226328.1 - 9 - 10 955 60

Gene Symbol
LOC100911453
Entrez Gene ID
100911453
Gene Name
methylosome protein 50-like
Gene Aliases
-
Chromosome Location
Chr.2: 208420203 - 208430283 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003749373.3 XP_003749421.1 9 - 10 1066 60

View Full Product Details