Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sox17
Entrez Gene ID
312936
Gene Name
SRY (sex determining region Y)-box 17
Gene Aliases
-
Chromosome Location
Chr.5: 14890309 - 14895907 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107902.1 NP_001101372.1 2 - 3 223 91

View Full Product Details