Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zmpste24
Entrez Gene ID
313564
Gene Name
zinc metallopeptidase STE24
Gene Aliases
-
Chromosome Location
Chr.5: 139983680 - 140015541 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107974.1 NP_001101444.1 1 - 2 445 71

View Full Product Details