Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gpatch3
Entrez Gene ID
362615
Gene Name
G patch domain containing 3
Gene Aliases
Gpatc3, RGD1311216
Chromosome Location
Chr.5: 151796456 - 151806225 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_342933.6 XP_342934.3 1 - 2 819 71

View Full Product Details