The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025696.1 | NP_001020867.2 | 12 - 13 | 1630 | 69 |
GenBank mRNA | BC098795.1 | - | 12 - 13 | 1630 | 69 |