Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zim1
Entrez Gene ID
308322
Gene Name
zinc finger, imprinted 1
Gene Aliases
-
Chromosome Location
Chr.1: 70244884 - 70266709 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107473.1 NP_001100943.1 5 - 6 562 91

View Full Product Details