Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
St6galnac1
Entrez Gene ID
287920
Gene Name
ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 1
Gene Aliases
Siat7A
Chromosome Location
Chr.10: 105713305 - 105723700 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105859.1 NP_001099329.1 2 - 3 716 90

View Full Product Details