Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr24
Entrez Gene ID
360497
Gene Name
WD repeat domain 24
Gene Aliases
RGD1304803
Chromosome Location
Chr.10: 15189772 - 15195860 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191084.1 NP_001178013.1 2 - 3 883 85
XM_008767587.1 XP_008765809.1 2 - 3 2189 85

View Full Product Details