The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105819.1 | NP_001099289.1 | 8 - 9 | 1196 | 113 |
XM_006246980.2 | XP_006247042.1 | 7 - 8 | 1233 | 113 | |
XM_006246981.1 | XP_006247043.1 | 8 - 9 | 1922 | 113 | |
XM_008767963.1 | XP_008766185.1 | 7 - 8 | 1033 | 113 |