Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Noxo1
Entrez Gene ID
302976
Gene Name
NADPH oxidase organizer 1
Gene Aliases
-
Chromosome Location
Chr.10: 14056416 - 14066918 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106986.1 NP_001100456.1 5 - 6 889 82
XM_006245948.1 XP_006246010.1 5 - 6 514 82
XM_006245949.1 XP_006246011.1 4 - 5 901 82
XM_006245950.2 XP_006246012.1 4 - 5 901 82
XM_008767566.1 XP_008765788.1 4 - 5 901 82

View Full Product Details