Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxw11
Entrez Gene ID
303024
Gene Name
F-box and WD repeat domain containing 11
Gene Aliases
-
Chromosome Location
Chr.10: 17542374 - 17640718 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106993.1 NP_001100463.1 3 - 4 308 97

View Full Product Details