Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mslnl
Entrez Gene ID
363554
Gene Name
mesothelin-like
Gene Aliases
RGD1561732
Chromosome Location
Chr.10: 15105281 - 15119427 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008767615.1 XP_008765837.1 10 - 11 3229 64

View Full Product Details