The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107017.2 | NP_001100487.1 | 3 - 4 | 381 | 58 |
GenBank mRNA | BC162047.1 | - | 3 - 4 | 381 | 58 |