Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ggact
Entrez Gene ID
290500
Gene Name
gamma-glutamylamine cyclotransferase
Gene Aliases
A2ld1, RGD1304748
Chromosome Location
Chr.15: 109307683 - 109336029 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006252521.2 XP_006252583.1 1 - 2 263 99

View Full Product Details