Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Asxl1
Entrez Gene ID
311553
Gene Name
additional sex combs like 1
Gene Aliases
RGD1561878
Chromosome Location
Chr.3: 148832231 - 148902356 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008762396.1 XP_008760618.1 8 - 9 6410 64

View Full Product Details