Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bpifb4
Entrez Gene ID
499925
Gene Name
BPI fold containing family B, member 4
Gene Aliases
Lplunc4, RY2G5
Chromosome Location
Chr.3: 149424392 - 149449350 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109209.2 NP_001102679.2 8 - 9 1154 149
GenBank mRNA X60660.1 - 6 - 7 713 149

View Full Product Details