Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rgr
Entrez Gene ID
306307
Gene Name
retinal G protein coupled receptor
Gene Aliases
-
Chromosome Location
Chr.16: 14286323 - 14300951 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107299.1 NP_001100769.1 5 - 6 670 70
XM_008771101.1 XP_008769323.1 5 - 6 658 70

View Full Product Details