The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011936.1 | NP_001011936.1 | 3 - 4 | 1025 | 72 |
GenBank mRNA | BC085857.1 | - | 3 - 4 | 1025 | 72 |