Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfand1
Entrez Gene ID
361917
Gene Name
zinc finger, AN1-type domain 1
Gene Aliases
RGD1309519
Chromosome Location
Chr.2: 93656918 - 93666745 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_342213.6 XP_342214.4 5 - 6 390 74
GenBank mRNA FQ229099.1 - 5 - 6 376 74

View Full Product Details