Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Comtd1
Entrez Gene ID
305685
Gene Name
catechol-O-methyltransferase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.15: 2631529 - 2634640 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107249.1 NP_001100719.1 6 - 7 699 132

View Full Product Details