Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sytl5
Entrez Gene ID
302538
Gene Name
synaptotagmin-like 5
Gene Aliases
-
Chromosome Location
Chr.X: 14017405 - 14109592 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178333.1 NP_848016.1 12 - 13 1504 82
XM_008773069.1 XP_008771291.1 14 - 15 2834 82
XM_008773070.1 XP_008771292.1 14 - 15 2825 82
GenBank mRNA AB098162.1 - 12 - 13 1504 82

View Full Product Details