The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001047849.1 | NP_001041314.1 | 12 - 13 | 2188 | 82 |
GenBank mRNA | BC082092.1 | - | 9 - 10 | 1034 | 82 |
BC161809.1 | - | 11 - 12 | 1474 | 82 | |
DQ480750.1 | - | 12 - 13 | 2188 | 82 |