The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107838.1 | NP_001101308.1 | 5 - 6 | 569 | 71 |
GenBank mRNA | BC168896.1 | - | 5 - 6 | 549 | 71 |