Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lcn10
Entrez Gene ID
499756
Gene Name
lipocalin 10
Gene Aliases
RGD1564362
Chromosome Location
Chr.3: 2894702 - 2898255 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128137.1 NP_001121609.1 3 - 4 336 74
GenBank mRNA DQ537494.1 - 3 - 4 336 74

View Full Product Details