Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr5
Entrez Gene ID
362093
Gene Name
WD repeat domain 5
Gene Aliases
-
Chromosome Location
Chr.3: 6061892 - 6080724 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039034.1 NP_001034123.1 10 - 11 1068 71
XM_006233831.2 XP_006233893.1 11 - 12 1208 71
GenBank mRNA BC100156.1 - 10 - 11 1068 71

View Full Product Details