Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc2a6
Entrez Gene ID
296600
Gene Name
solute carrier family 2 (facilitated glucose transporter), member 6
Gene Aliases
-
Chromosome Location
Chr.3: 5567729 - 5575136 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106562.1 NP_001100032.1 2 - 3 339 96
XM_006233757.2 XP_006233819.1 2 - 3 334 96
XM_008761634.1 XP_008759856.1 2 - 3 334 96

View Full Product Details