The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107193.1 | NP_001100663.1 | 1 - 2 | 401 | 87 |
XM_008769651.1 | XP_008767873.1 | 1 - 2 | 1077 | 87 | |
XM_008769652.1 | XP_008767874.1 | 2 - 3 | 438 | 87 |