Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbx19
Entrez Gene ID
304935
Gene Name
T-box 19
Gene Aliases
-
Chromosome Location
Chr.13: 83403263 - 83426317 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107193.1 NP_001100663.1 6 - 7 1194 70
XM_008769651.1 XP_008767873.1 7 - 8 1932 70
XM_008769652.1 XP_008767874.1 8 - 9 1293 70

View Full Product Details