Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fmo13
Entrez Gene ID
289193
Gene Name
flavin-containing monooxygenase 13
Gene Aliases
-
Chromosome Location
Chr.13: 84682409 - 84694575 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008769727.1 XP_008767949.1 1 - 2 132 131

View Full Product Details